missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Adjuvants

Product Code. 11762934
Click to view available options
Product Type:
Complete H37Ra
Freund's Complete Adjuvant
Freund's Incomplete Adjuvant
Mycobacterium butyricum
Mycobacterium tuberculosis H37Ra
Quantity:
6 x 100 mg
6 x 10 mL
Unit Size:
600mg
60mL
Pack of 6
This item is not returnable. View return policy
Not intended for human or therapeutic use.

Product Code. 11762934

Brand: BD 231131

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Not intended for human or therapeutic use.

Used to prepare antigen-adjuvant emulsions. BD™ Difco™ Adjuvants are used in immunological studies with laboratory animals. (Not intended for human or therapeutic use.)

  • Adjuvants are mixed with aqueous antigen solution or suspension
  • Once injected, they provide a pool of emulsified antigen at the injection site and allow the antigen to be released over an extended period of time
  • With adjuvants, the immune response to antigens (especially soluble antigens) is enhanced and prolonged
  • Use Incomplete Freund adjuvant to avoid the use of acid-fast bacilli; however, the antibody response is not as marked as with the complete adjuvant
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in cool, dry conditions in well sealed containers, away from oxidizing agents.
For Use With (Application) Immunological studies with laboratory animals
Formulation Suspension of 10mg M. tuberculosis in a mixture of paraffin oil and an emulsifying agent
Quantity 6 x 10 mL
Product Type Complete H37Ra
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.