missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Adjuvants
Click to view available options
Product Type:
Complete H37Ra
Freund's Complete Adjuvant
Freund's Incomplete Adjuvant
Mycobacterium butyricum
Mycobacterium tuberculosis H37Ra
Quantity:
6 x 100 mg
6 x 10 mL
Unit Size:
600mg
60mL
Pack of 6
Description
- Adjuvants are mixed with aqueous antigen solution or suspension
- Once injected, they provide a pool of emulsified antigen at the injection site and allow the antigen to be released over an extended period of time
- With adjuvants, the immune response to antigens (especially soluble antigens) is enhanced and prolonged
- Use Incomplete Freund adjuvant to avoid the use of acid-fast bacilli; however, the antibody response is not as marked as with the complete adjuvant
Specifications
Specifications
| Content And Storage | Store in cool, dry conditions in well sealed containers, away from oxidizing agents. |
| For Use With (Application) | Immunological studies with laboratory animals |
| Formulation | Killed M. butyricum for use in adjuvants |
| Quantity | 6 x 100 mg |
| Product Type | Mycobacterium butyricum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction