missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Antisera for Serological Confirmation of E. coli O157:H7

E. ColiO Antiserum O157 and E. Coli H Antiserum H7 are used for identifying Escherichia coli O157:H7

Brand:  BD 221591


 View more versions of this product

Product Code. 11748323

  • £201.00 / 3mL
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

E. coli O157:H7 is an enteric pathogen that requires only a low inoculum to cause disease. Transmission is usually via food items. Serotyping of the entero-hemorrhagic E. coli is useful in the epidemiological documentation of the spread of a particular strain in a foodborne outbreak. At the time of use, test both positive and negative control cultures to check performance of the antiserum.

  • Product type: antigens and antisera
  • Groupings: D through K Products
  • Type: antisera
  • Sub type: E.coli - Lyophilized, re-hydrate to 3mL
  • Shelf Life: 1095 days
  • Label has Natural Rubber Latex statement

Certifications

CE marked

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CE Marked
Refrigerate, store at 2°C to 8°C
Antiserum
For tube test for serological confirmation of E. coli O157:H7
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • BD Difco E. Coli H Antiserum H7, 3 mL

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : sodium azide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers