missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Bordetella spp. Antigen and Antisera

Quick reactive Gold standard antisera-typing panels for Bordetella Pertusis antigen and Bordetella Parapertusis antisera

Brand:  BD 225851


 View more versions of this product

Product Code. 11702844

  • £238.00 / 5mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

This item is not returnable. View return policy

Description

Description

  • Product type:     Antigens and Antisera
  • Groupings: A through B products
Bordetella Pertussis Antigen
  • Sub type: Quality control antigens
  • Type: Antigens
  • Quantity: 5mL
  • Shelf Life: 730 days
  • Positive Control for Bordetella pertussis antiserum
Bordetella Parapertussis Antiserum
  • Subtype: Bordetella Antisera
  • Type: Antisera
  • Lyophilized, rehydrate to 1mL
  • Shelf Life: 965 days
  • Rabbit serum for slide agglutination

Certifications

CE marked

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CE Marked
Antisera
5 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • BD Difco Bordetella Pertussis Antigen, 5 mL

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : sodium azide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers