missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Gonococcal (GC) Medium Base

Product Code. 11752904
Click to view available options
:
Gonococcal (GC) Medium Base; 500g
This item is not returnable. View return policy

Product Code. 11752904

Brand: BD 228950

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For various additives in isolating and cultivating Neisseria gonorrhoeae and other fastidious microorganisms. BD Difco™ Dehydrated Culture Media: Gonococcal (GC) Medium Base is part of a large selection of dehydrated media.

  • Dehydrated
  • Culture Media
  • Gonococcal (GC) Medium Base
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage <30°C
Form Dehydrated
Media Type GC Medium Base
Product Type Microbiology Media
Formulation Proteose Peptone No. 3 15g/L, Corn Starch 1g/L, Dipotassium Phosphate 4g/L, Monopotassium Phosphate 1g/L, Sodium Chloride 5g/L, Agar 10g/L
For Use With (Application) For various additives in isolating and cultivating Neisseria gonorrhoeae and other fastidious microorganisms
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.