Learn More
BD Difco™ Lactobacilli MRS Broth
Recommended for use in the isolation, enumeration and cultivation of Lactobacillus species. BD™ Difco™ Lactobacilli MRS Broth supports luxuriant growth of all lactobacilli from oral, fecal, dairy and other sources. Supplied in a convenient, easy-to-use dehydrated powder.
Brand: BD 288110
412.94 GBP valid until 2024-12-31
Use promo code "21946" to get your promotional price.
Additional Details : Weight : 0.80000kg
Description
Difco™ Lactobacilli MRS Broth powder contains peptone and dextrose. These ingredients supply nitrogen, carbon and other elements necessary for growth. Polysorbate 80, acetate, magnesium and manganese provide growth factors for culturing a variety of lactobacilli. The above ingredients may inhibit the growth of some organisms other than lactobacilli.
- Based on the formulations of deMan, Rogosa and Sharpe
- Samples can be inoculated directly into Lactobacilli MRS Broth
- Produces a good cultural response with: Lactobacillus delbrueckii (subsp. lactis) 7830, Lactobacillus fermentum 9338 and Lactobacillus johnsonii 11506
- pH (5.5% solution, 25°C): 6.5 ± 0.2
Specifications
COMPF, SMD | |
2°C to 8°C | |
MRS Broth | |
Proteose Peptone No. 3 10g/L, Beef extract 10g/L, Yeast extract 5g/L, Dextrose 20g/L, Polysorbate 80 1g/L, Ammonium Citrate 2g/L, Sodium Acetate 5g/L, Magnesium Sulfate 0.1g/L, Manganese Sulfate 0.05g/L, Dipotassium Phosphate 2g/L | |
Pail |
Tan | |
Dehydrated Powder | |
Microbiology Media | |
Cultivation of lactobacilli | |
2 kg |