missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: LB Agar, Miller (Luria-Bertani)
Propagate and maintain E. coli for molecular biology. BD Difco™ Dehydrated Culture Media: LB Agar, Miller (Luria-Bertani) supplied in dehydrated form for the convenience of the laboratorian and bioprocessing specialist.
£107.00 - £386.00
Specifications
Certifications/Compliance | CCAM |
---|---|
Content And Storage | <30°C |
Form | Dehydrated |
Media Type | LB Agar, Miller |
Product Type | Agar |
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
11748902
|
BD
244510 |
Pail | 2000 g |
£386.00
2kg |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||||
11758902
|
BD
244520 |
Bottle | 500 g |
£107.00
500g |
In Stock
Log in to see stock. |
||||
Description
- Contains 10g/L NaCl
- For propagation of E. coli
- Wide range of molecular biology applications
Specifications
CCAM | |
Dehydrated | |
Agar | |
Propagation and maintenance of Escherichia coli for molecular biology |
<30°C | |
LB Agar, Miller | |
Tryptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L, Agar 15g/L |