missing translation for 'onlineSavingsMsg'
Learn More
BD Difco™ Dehydrated Culture Media: LB Broth, Miller (Luria-Bertani) Product Code.: 11768902

BD Difco™ Dehydrated Culture Media: LB Broth, Miller (Luria-Bertani)

Product Code. p-4869129
2000 g, Pail, 2kg
Click to view available options
Quantity:
500 g
2000 g
Packaging Type:
Bottle
Pail
Unit Size:
2kg
500g
This item is not returnable. View return policy

Product Code. 11768902

Brand: BD 244610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Propagate and maintain E. coli for molecular biology. BD Difco™ Dehydrated Culture Media: LB Broth, Miller (Luria-Bertani) supplied in dehydrated form for the convenience of the laboratorian and bioprocessing specialist.

 

  • Contains 10g/L NaCl
  • For propagation of E. coli
  • Wide range of molecular biology applications

 

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CCAM
Content And Storage Refrigerate
Form Dehydrated
Media Type LB Broth, Miller
Product Type Microbiology Media
Formulation Tryptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L
For Use With (Application) Propagation and maintenance of Escherichia coli for molecular biology
Packaging Type Pail
Quantity 2000 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BD Difco™ Dehydrated Culture Media: LB Broth, Miller (Luria-Bertani) >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.