Learn More
BD Difco™ Dehydrated Culture Media: Leptospira Medium Base EMJH
Use with Leptospira Enrichment EMJH to cultivate and maintain Leptospira
Brand: BD 279410
Additional Details : Weight : 0.80000kg
Description
Applications:
For evaluation, maintenance, selective enrichment, differentiation, isolation, cultivation, detection, propagation of bacteria
Specifications
SMWW | |
Dehydrated | |
Microbiology Media | |
Used with Leptospira enrichment EMJH to cultivate and maintain Leptospira | |
500 g |
Refrigerate | |
Leptospira Medium Base | |
Disodium Phosphate 1g/L, Monopotassium Phosphate 0.3g/L, Sodium Chloride 1g/L, Ammonium Chloride 0.25g/L, Thiamine 5mg/L | |
Bottle |
Safety and Handling
- leptosphira
Signal Word
- Warning
Hazard Category
- Specific target organ toxicity Category 3
Hazard Statement
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P405-Store locked up.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ammonium chloride