missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Middlebrook 7H9 Broth
Click to view available options
:
Middlebrook 7H9 Broth; 500g
Description
- Use with: Tween™ 80 or Glycerol and Middlebrook ADC
- Dehydrated
- Culture Media
- Middlebrook 7H9 Broth
Order Info
See our web site or the Fisher Scientific™ HealthCare catalog for product information.
Specifications
Specifications
| Certifications/Compliance | AOAC, SMWW |
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | Middlebrook 7H9 Broth |
| Product Type | Microbiology Media |
| Formulation | Ammonium Sulfate 0.5g/L, L-Glutamic Acid 0.5g/L, Sodium Citrate 0.1g/L, Pyridoxine 1mg/L, Biotin 0.5mg/L, Disodium Phosphate 2.5g/L, Monopotassium Phosphate 1g/L, Ferric Ammonium Citrate 0.04g/L, Magnesium Sulfate 0.05g/L, Calcium Chloride 0.5mg/L, Zinc Sulfate 1mg/L, Copper Sulfate 1mg/L |
| For Use With (Application) | Cultivation of Mycobacteria |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction