Learn More
BD Difco™ Dehydrated Culture Media: Modified Letheen Broth (AOAC)
For determining antibiotic potency by the microbiological assay technique
Brand:  BD 263010
Product Code. 11709052
  • £194.00 / 500g
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Applications:
For evaluation, maintenance, selective enrichment, differentiation, isolation, cultivation, detection, propagation of bacteria
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BAM
Dehydrated
Microbiology Media
Microbiological evaluation of cosmetics - inactivates preservative agents
500 g
2°C to 8°C
Letheen Broth
Letheen broth 25.7g/L, Tryptone 5g/L, Proteose Peptone No. 3 10g/L, Yeast extract 2g/L, Sodium Bisulfite 0.1g/L
Bottle
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BD Difco™ Dehydrated Culture Media: Modified Letheen Broth (AOAC) > Letheen Broth; 500g