missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Seven H11 Agar Base
For isolation of Mycobacterium tuberculosis
Brand: BD 212203
Additional Details : Weight : 0.50000kg
Specifications
ISO/DIS, 11014 | |
Dehydrated | |
Agar | |
Isolation of Mycobacterium tuberculosis | |
500 g |
25°C | |
Seven H11 Agar Base | |
Pancreatic digest of Casein 1g/L, Monosodium Glutamate 0.5g/L, Sodium Citrate 0.4g/L, Pyridoxine 1mg/L, Biotin 0.5mg/L, Ferric Ammonium Citrate 0.04g/L, Ammonium Sulfate 0.5g/L, Disodium Phosphate 1.5g/L, Monopotassium Phosphate 1.5g/L, Magnesium Sulfate | |
Bottle |