Learn More
BD Difco™ Agar, Technical
Used in preparing microbiological culture media. BD™ Difco™ Agar, Technical is a solidifying agent with wider quality control parameters than other bacteriological agars. However, the solubility, gelation temperature and solidity are carefully monitored to permit its use.
£214.00 - £626.00
Specifications
Certifications/Compliance | ISO/DIS, 11014 |
---|---|
Color | Very light to medium beige |
Concentration | 1.5% |
Content And Storage | <30°C |
Additive | Technical Grade Agar |
Product Code | Brand | Quantity | Packaging Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging Type | Price | Quantity & Availability | ||||
11783523
|
BD
281210 |
2 kg | Container |
£626.00
2kg |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
11793523
|
BD
281230 |
500 g | Bottle |
£214.00
500g |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
Description
Technical Agar is suitable for many bacteriological applications. This agar is not highly processed, has broader technical specifications than other agars and is not recommended for growth of fastidious organisms. It shows good cultural response (in nutrient broth) to Escherichia coli 25922 and Staphylococcus aureus 25923.
- Loss on Drying (LOD): ≤20%
- Ash(%): ≤6.5%
- Calcium μg/g (ppm): ≤3,000 ppm
- Magnesium μg/g (ppm): ≤1,300 ppm
- Melting Point: ≤85°C
- Gelation Point: 32–39°C
- Gel Strength: 613g/cm3
- pH (1.5% solution, 25°C): 5.9
- Spore count: 4,300 CFU/g
- Standard plate count: 2,725 CFU/g
- Inorganics: Calcium 0.110%, Chloride 0.172%, Cobalt <0.001%, Copper <0.001%, Iron 0.002%, Lead <0.001%, Magnesium 0.093%, Manganese <0.001%, Nitrate <0.005%, Phosphate 0.015%, Potassium 0.124%, Sodium 0.932%, Sulfate 0.367%, Sulfur 0.646%, Tin <0.001%, Zinc <0.001%
Agar is a phycocolloid extracted from a group of red-purple marine algae (Class Rhodophyceae). Agar is the gelling agent of choice because it remains firm at growth temperatures for many pathogens. It is also resistant to a breakdown by bacterial enzymes. Agar melts at a different temperature than that at which it solidifies, a property known as hysteresis. It is generally resistant to shear forces; however, different agars may have different gel strengths. Agar is typically used in a final concentration of 1–2% for solidifying culture media.
Specifications
ISO/DIS, 11014 | |
1.5% | |
Technical Grade Agar | |
Technical |
Very light to medium beige | |
<30°C | |
Dehydrated Powder | |
Bacteriological culture media |