missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Yeast Mold Broth

For cultivation of yeasts, molds and other aciduric microorganisms. BD Difco™ Dehydrated Culture Media: Yeast Mold Broth is part of a large selection of dehydrated media.

Brand:  BD 271120

Additional Details : Weight : 0.80000kg

Product Code. 11733473

  • £283.00 / 500g
Estimated Shipment: 23-07-2024
to see stock.
Add to basket

Description

Description

  • Dehydrated
  • Culture Media
  • Yeast Mold Broth

Order Info

Prices are for units of one each unless otherwise specified.

Specifications

Specifications

<30°C
Yeast Mold Broth
Yeast extract 3g/L, Malt extract 3g/L, Peptone 5g/L, Dextrose 10g/L
Bottle
Dehydrated
Microbiology Media
Cultivation of Yeasts, molds and other Aciduric microorganisms
500 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers