missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BD™ Difco™ Dextrose Glucose, Anhydrous
Used with microbiological culture media. BD™ Difco™ Dextrose Glucose, Anhydrous is a form of D(+)-Glucose. The dehydrated form provides greater convenience for the laboratorian and bioprocessing specialist.
£65.40
Specifications
Certifications/Compliance | ISO/DIS, 11014 |
---|---|
Color | Whitish |
Content And Storage | <30°C |
Additive | Dextrose Glucose |
Form | Anhydrous Solid |
Description
D-Glucose is a simple sugar that circulates in the blood of animals as blood sugar, and is made in plants during photosynthesis. It is an important source of power for cellular respiration. Glucose is stored as as a starch in plants and as glycogen in animals.
- CAS: 50-99-7
- EC No.: 200-075-1
- Chemical formula: C6H12O6
- Molar mass: 180.16g/mol
- Synonym: Blood Sugar
Specifications
ISO/DIS, 11014 | |
<30°C | |
Anhydrous Solid |
Whitish | |
Dextrose Glucose | |
Microbiological culture media |