missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Fluorescent Antibody Reagents

Reagents conjugated with FITC or Rhodamine to make reactions visible under fluorescent light

£19.35 - £1125.06

Specifications

Quantity 5 mL
For Use With (Application) Direct fluorescent antibody technique for the identification of Bordetella pertussis and Bordetella parapertussis
View More Specs

Products 3
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
11750234
View Documents
BD
223591
FA Bordetella Pertussis
£19.35
Each
Estimated Shipment: 28-02-2025
to see stock.
Add to basket
 
11722784
View Documents
BD
223591
FA Bordetella Pertussis This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11732784
View Documents
BD
223781
FA Bordetella Parapertussis This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Reagents conjugated with FITC or Rhodamine to make reactions visible under fluorescent light
  • For use in direct fluorescent identification

Order Info

In 5mL vials.

Specifications

Specifications

5 mL
Direct fluorescent antibody technique for the identification of Bordetella pertussis and Bordetella parapertussis
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.