missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ HYcheck™ Contact Slides: Disinfection Control
Used to assess the microbiological contamination of surfaces or fluids. BD Difco™ HYcheck™ Contact Slides: Disinfection Control are designed for environmental testing.
Brand: BD 290002
Additional Details : Weight : 0.80000kg
Description
- Designed for environmental testing
- Double sided with Tryptic Soy and D/E Neutralizing Agar
- Hinged paddle bends for easy sampling
Order Info
Ten units per pack.
Specifications
Powder | |
Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 15g/L, Pancreatic digest of Casein 5g/L, Yeast extract 2.5g/L, Dextrose 10g/L, Sodium Thioglycollate 1g/L, Sodium Thiosulfate 6g/L, Sodium Bisulfite 2.5g/L, | |
Slide |
United States | |
Assess the microbiological contamination of surfaces or fluids | |
10 U |