missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Haemophilus influenzae Serotyping Antisera

Use in slide agglutination tests for serotyping Haemophilus influenzae

Brand:  BD 222371


 View more versions of this product

Product Code. 11722734

  • £647.00 / 1mL
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket

Alert:

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

This item is not returnable. View return policy

Description

Description

  • Lyophilized
  • Rehydrate to 1mL
  • Type: Antisera
  • Shelf Life: 965 days

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Refrigerate, store at 2°C to 8°C
Haemophilus influenzae Antiserum Poly
Antiserum
Use in slide agglutination tests for serotyping Haemophilus influenzae
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501-Dispose of contents/container to in accordance with all local/regional/national/internationalregulations.

Supplemental information
  • MIXTURE LIST-Contains : Sodium Azide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers