missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Haemophilus influenzae Serotyping Antisera

Use in slide agglutination tests for serotyping Haemophilus influenzae

Brand:  BD 222361


 View more versions of this product

Product Code. 11788333

  • £551.00 / 1mL
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket

Alert:

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

This item is not returnable. View return policy

Description

Description

  • Lyophilized
  • Rehydrate to 1mL
  • Type: Antisera
  • Shelf Life: 965 days

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Refrigerate, store at 2°C to 8°C
Haemophilus influenzae Antiserum Type b
Antiserum
Use in slide agglutination tests for serotyping Haemophilus influenzae
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Vial H Influenzae Antiserum Type B

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P270-Do not eat, drink or smoke when using this product.
  • P264-Wash thoroughly after handling.
  • P330-Rinse mouth.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P501-Dispose of contents/container to in accordance with all local/regional/national/internationalregulations.

Supplemental information
  • MIXTURE LIST-Contains : sodium azide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.