Learn More
BD Difco™ Neisseria Meningitidis Antisera
For serotyping Neisseria meningitidis in slide agglutination tests
Brand: BD 222531
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Description
- Stable, lyophilized, polyclonal rabbit antisera
- Contains 0.1% sodium azide as preservative
Specifications
For in vitro diagnostic use | |
Neisseria meningitidis Antiserum Group W135 | |
Antiserum | |
1 mL |
Safety and Handling
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P270-Do not eat, drink or smoke when using this product.
- P264-Wash thoroughly after handling.
- P330-Rinse mouth.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : sodium azide
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For in vitro diagnostic use.