Learn More
BD BD™ Difco™ R2A Agar
Intended for use with stressed and chlorine-tolerant bacteria. BD™ Difco™ R2A Agar is a dehydrated, low-nutrient medium for enumerating and cultivating compromised bacteria and other heterotrophic organisms. It was developed for bacteriological plate counts of treated potable water.
£229.00 - £530.00
Specifications
Certifications/Compliance | COMPF, EPA, SMWW |
---|---|
Color | Light beige |
Form | Dehydrated (free-flowing, homogenous) |
Media Type | R2A Agar |
Product Type | Agar |
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
11798273
|
BD
218263 |
Bottle | 500 g |
£229.00
500g |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
17392301
|
BD
218261 |
Pail | 2 kg |
£530.00
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||||
Description
R2A Agar was developed for bacteriological plate counts of treated potable water. A low nutrient medium, such as R2A Agar, in combination with a lower incubation temperature and longer incubation time stimulates the growth of stressed and chlorine-tolerant bacteria.
Nutritionally-rich media, such as Plate Count Agar, support the growth of fast-growing bacteria but may suppress slow growing or stressed bacteria found in treated water. When compared with nutritionally rich media, R2A Agar has been reported to improve the recovery of stressed and chlorine-tolerant bacteria from drinking water systems.
- Recommended in standard methods for pour plate, spread plate and membrane filter methods for heterotrophic plate counts.
- Intended for use only with compromised bacteria found in treated potable water.
- Yeast extract provides a source of trace elements and vitamins.
- Peptone and casamino acids provide nitrogen, vitamins, amino acids, carbon and minerals.
- Dextrose serves as a carbon source.
- Soluble starch aids in the recovery of injured organisms by absorbing toxic metabolic by-products.
- Sodium pyruvate increases the recovery of stressed cells.
- Potassium phosphate is used to balance the pH and provide phosphate.
- Magnesium sulfate is a source of divalent cations and sulfate.
- Agar is the solidifying agent.
Order Info
Prices are for units of one each unless otherwise specified.
Certifications
COMPF, EPA, SMWW
Specifications
COMPF, EPA, SMWW | |
Dehydrated (free-flowing, homogenous) | |
Agar | |
Low nutrient medium for enumeration and cultivation of bacteria from potable Water |
Light beige | |
R2A Agar | |
Yeast extract 0.5g/L, Proteose Peptone No. 3 0.5g/L, Casamino Acids 0.5g/L, Dextrose 0.5g/L, Soluble Starch 0.5g/L, Sodium Pyruvate 0.3g/L, Dipotassium Phosphate 0.3g/L, Magnesium Sulfate 0.05g/L, Agar 15g/L |