Learn More
BD Difco™ Salmonella H Antiserum G Complex
Qualitative tube agglutination tests for serological differentiation of Salmonella
Brand: BD 222691
Alert:
The products contains sodium azide which is toxic by inhalation, by skin contact, and if swallowed.Sodium azide may react with lead and copper plumbing to form highly explosive metal azides. On disposal, flush with large volume of water to prevent azide build-up. After contact with skin, wash immediately with plenty of water.Packaging contains dry natural rubber.
Description
- Lyophilized and polyclonal rabbit antisera
- Absorbed or unabsorbed specifically for either phase 1 or phase 2 antigens
- Perform between 150-1500 tests
- Identification of Salmonella by flagellar (H) antigens
- Sodium azide (0.2%) used as preservative
Specifications
Antiserum | |
3 mL |
Safety and Handling
- Vial Sal H Antiserum G Complex
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Sodium Azide
Your input is important to us. Please complete this form to provide feedback related to the content on this product.