missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Salmonella H Antiserum Poly D, Factors z35, z36, z37, z38, z39, z41, z 42

Qualitative tube agglutination tests for serological differentiation of Salmonella

Brand:  BD 225421

Additional Details : Weight : 0.00300kg

Product Code. 11702824

  • £478.00 / 3mL
Estimated Shipment: 16-08-2024
to see stock.
Add to basket

Alert:

The products contains sodium azide which is toxic by inhalation, by skin contact, and if swallowed.Sodium azide may react with lead and copper plumbing to form highly explosive metal azides. On disposal, flush with large volume of water to prevent azide build-up. After contact with skin, wash immediately with plenty of water.Packaging contains dry natural rubber.


Description

Description

  • Lyophilized and polyclonal rabbit antisera
  • Absorbed or unabsorbed specifically for either phase 1 or phase 2 antigens
  • Perform between 150-1500 tests
  • Identification of Salmonella by flagellar (H) antigens
  • Sodium azide (0.2%) used as preservative

Specifications

Specifications

Antiserum
3 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers