missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Salmonella H Antiserum Single Factor 5

Product Code. 11772794
Click to view available options
Quantity:
3 mL
Unit Size:
3mL
This item is not returnable. View return policy
The products contains sodium azide which is toxic by inhalation, by skin contact, and if swallowed.Sodium azide may react with lead and copper plumbing to form highly explosive metal azides. On disposal, flush with large volume of water to prevent azide build-up. After contact with skin, wash immediately with plenty of water.Packaging contains dry natural rubber.

Product Code. 11772794

Brand: BD 224751

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
The products contains sodium azide which is toxic by inhalation, by skin contact, and if swallowed.Sodium azide may react with lead and copper plumbing to form highly explosive metal azides. On disposal, flush with large volume of water to prevent azide build-up. After contact with skin, wash immediately with plenty of water.Packaging contains dry natural rubber.

Qualitative tube agglutination tests for serological differentiation of Salmonella

  • Lyophilized and polyclonal rabbit antisera
  • Absorbed or unabsorbed specifically for either phase 1 or phase 2 antigens
  • Perform between 150-1500 tests
  • Identification of Salmonella by flagellar (H) antigens
  • Sodium azide (0.2%) used as preservative

TRUSTED_SUSTAINABILITY

Specifications

Product Type Antiserum
Quantity 3 mL
compliance-icons
Product Identifier
  • Vial Sal H Antiserum Single Factor 5 3Ml
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Sodium Azide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.