missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Salmonella O Antisera and Quality Control Antigens

Brand:  BD 228151


 View more versions of this product

Product Code. 11702884

  • £140.00 / 3mL
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket

Alert:

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

This item is not returnable. View return policy

Description

Description

Salmonella species cause a variety of human diseases called salmonelloses. The range of disease is from mild self-limiting gastroenteritis to more severe forms, possibly with bacteremia or typhoid fever, which can be life threatening. Hence diagnosis and identification of correct serotype is essential. These antisera aid in detecting such serotypes and the quality control antigens check their performance.

  • Lyophilized, polyclonal rabbit antisera
  • Polyvalent, specific for somatic antigens
  • Cross-reactions are possible because serogroups may share non-major group antigens
  • When properly rehydrated and used as recommended, each vial contains sufficient reagent for 60 tests
  • For in vitro diagnostic use
  • containing approximately 0.2% sodium azide as a preservative

TRUSTED_SUSTAINABILITY
Specifications

Specifications

For in vitro diagnostic use
Antiserum
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Vial Sal O Antiserum Factors 4& 5 Grp B

Signal Word
  • Warning

Clasificación
  • Acute toxicity Category 4

Indicaciones de peligro
  • H302-Harmful if swallowed.

Consejos de prudencia
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Otros peligros
  • MIXTURE LIST-Contains : Sodium Azide

SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales