missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Salmonella O Antisera and Quality Control Antigens
Click to view available options
Product Type:
Antiserum
Unit Size:
3mL
Each
Description
Salmonella species cause a variety of human diseases called salmonelloses. The range of disease is from mild self-limiting gastroenteritis to more severe forms, possibly with bacteremia or typhoid fever, which can be life threatening. Hence diagnosis and identification of correct serotype is essential. These antisera aid in detecting such serotypes and the quality control antigens check their performance.
- Lyophilized, polyclonal rabbit antisera
- Polyvalent, specific for somatic antigens
- Cross-reactions are possible because serogroups may share non-major group antigens
- When properly rehydrated and used as recommended, each vial contains sufficient reagent for 60 tests
- For in vitro diagnostic use
- containing approximately 0.2% sodium azide as a preservative
Specifications
Specifications
| Certifications/Compliance | For in vitro diagnostic use |
| Product Type | Antiserum |
Safety and Handling
Product Identifier
- Vial Sal O Antiserum Factor 9 Group D
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : sodium azide
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction