missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Salmonella O QC Antigens

Quick reactive Gold standard antisera-typing panels for Salmonella O antigen

Brand:  BD 221301

 View more versions of this product

Product Code. 11768313

  • £179.00 / 1mL
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket

Alert:

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

This item is not returnable. View return policy

Description

Description

  • Product type: Antigens and Antisera
  • Groupings: Q through S products
  • Sub type: Quality control antigens
  • Type: Antigens
  • Volume: 1mL
  • Shelf life: 730 days

Certifications

CE marked

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Antigen
Used in the quality control of Salmonella O Antisera Poly B (225351), Factor 7 (228161), Group C1 Factors 6 (240985), and Poly A-I & Vi (240993)
1 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers