missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Vibrio cholerae Antisera

Quick reactive Gold standard Vibrio cholerae Antisera

Brand:  BD 224321


 View more versions of this product

Product Code. 11742794

  • £85.00 / 3mL
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Product type: Antigens and Antisera
  • Groupings: U through Z Products
  • Sub type: Vibrio cholerae
  • Type: Antisera
  • Lyophilized, rehydrate to 3mL
  • Sufficient reagent for 20 slide tests
  • Available with 3 different antiserum types and O Antibodies
  • Shelf Life: 1095 days

Rabbit serum for slide agglutination test

Certifications

CE marked

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CE Marked
Refrigerate, store at 2°C to 8°C
Microbiology Media
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • BD Difco Vibrio Cholerae Antiserum Poly, 3 mL

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : sodium azide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers