Learn More
BD Disposable Syringes with Luer-Lok™ Tips
Unique integrated BD Luer-Lok™ tip syringe with clear barrel. BD Medical™ Disposable Syringes with Luer-Lok™ Tips are sterile and disposable.
£47.34 - £397.27
Specifications
Product Type Syringe
Disposable Single-use
Connection Type Luer Lock
Sterility Sterile
View More Specs
Products 3
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
12808130
View Documents
BD
BD309628
1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15880305
View Documents
BD
309628
1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10630694
View Documents
BD
309628
1 mL
£190.44
Pack of 100
Description
Description
Unique integrated BD Luer-Lok™ tip syringe with clear barrel. BD Medical™ Disposable Syringes with Luer-Lok™ Tips are sterile and disposable.
  • Sterile syringe for single use
  • Without needle
  • Clear barrel (polycarbonate or polypropylene) with bold scale markings
  • Tapered plunger rod for ease of aspiration
  • Positive plunger rod stop
  • Added BD Luer-Lok™ thread for increased secure connection
  • DNR (Latex) free
  • Shelf life: 1825 days
Specifications
Specifications
Syringe
Single-use
Luer Lock
Sterile
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
BD Disposable Syringes with Luer-Lok™ Tips