Learn More
BD DrySlide™ Reagent Slides
A convenient method for bacterial biochemical analysis. Self-contained, ready-to-use test slides consist of coated, 35mm film or impregnated paper, which operates as a carrier matrix for microbiological testing reagents.
Brand: BD 231747
This item is not returnable.
View return policy
Description
- One-step procedure —no need for rehydrating
- No thin paper strips to manipulate and no glass slides are involved
- Each individual slide contains four reaction areas
- DrySlide Indole is a 30-second, spot indole test that uses DMABA (5% p-dimethylaminobenzaldehyde) formulation to test for indole-producing bacteria
- DrySlide Nitrocefin is a qualitative test to detect the presence of bacteria that produces β-lactamase and meets NCCLS guidelines; each slide contains a reagent cassette impregnated with nitrocefin
- DrySlide Oxidase with each slide is film-coated with an oxidase reagent (N, N, N',N'-tetramethyl-p-phenylenediamine dihydrochloride) to test for the presence of cytochrome C oxidase activity in bacteria; a positive result produces a purple color within 20 seconds
- DrySlide PYR Kit has L-pyrrolidonyl-β-naphthylamide that is used for the rapid presumptive identification of Group D Enterococci and Strep Group A; includes 15 slides and 15, 0.5mL crush ampules
Order Info
Slides are individually wrapped and supplied in convenient dispenser packs
Specifications
PYR Kit | |
CE | |
Used for detecting hydrolysis of PYR in the rapid presumptive identification of group A streptococci and group D enterococci from pure culture |
15 Pack | |
Slide | |
15° to 30°C |