missing translation for 'onlineSavingsMsg'
Learn More

BD Vacutainer™ Eclipse™ Blood Collection Needle

Product Code. 12698805
Click to view available options
Gauge:
21 G
22 G
Includes:
Tube holder, safety shield
Color:
Black Hub
Green Hub
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 12698805

Brand: BD 368650

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Provides fast, easy needle-tip protection. BD Vacutainer™ Eclipse™ Blood Collection Needle is a multi-sample blood collection needle with an integrated safety shield that offers a simple, effective way to collect blood while reducing the possibility of needle stick injuries.

The BD Vacutainer™ Eclipse™ Blood Collection Needle features a safety shield that allows for one-handed activation to cover the needle immediately upon withdrawal from the vein and confirms proper activation with an audible click. Some options include a pre-attached tube holder that is ready to use right out of the package, with no assembly required. The integrated safety shield and holder maximize OSHA compliance by protecting clinicians from potential front and back-end needlestick injuries.

  • Integrated, single-use blood collection needle with Luer adapter
  • Intuitive and easy to use, so there is minimal change to your technique
  • Needle bevel up orientation toward the safety shield provides easy identification of bevel position
  • Safety shield is an integral part of the needle; it eliminates the need for secondary safety accessory device
  • Immediate one-handed safety feature activation reduces exposure time and doesn't require a hard surface for activation
  • Unique, double-locking system features an audible 'click' to ensure that the needle is shielded properly
  • Safety shield design allows hands to remain behind the needle at all times
  • Designed to help your institution comply with OSHA SHIB 10-15-03 (Disposal of Contaminated Needles and Blood Tube Holders Used for Phlebotomy)
TRUSTED_SUSTAINABILITY

Specifications

Product Type Blood Collection Needle
Sterility Sterile
Actuation Single-handed
Certifications/Compliance OSHA SHIB 10-15-03
Color Green Hub
Disposable Single-use
For Use With (Application) Blood Collection with reduced possibility of needlestick injuries
Gauge 21 G
Includes Tube holder, safety shield
Latex Free Yes
Length (English) 1.25 in.
Length (Metric) 3.17 cm
Style Bevel Point
Thickness (Wall) Thin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.