missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD ELISPOT Streptavidin-HRP for ELISPOT
Recommended for use with BD ELISPOT Pairs
Brand: BD 557630
Additional Details : Weight : 0.01000kg
Description
- Routinely tested by ELISPOT
- Storage buffer: Aqueous buffered solution containing BSA and ProClin™ 150 as preservative
Specifications
Light Brown | |
1 mL | |
Liquid |
HRP | |
4°C | |
ELISPOT |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HRP Streptavidin for ELISPOT
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P363-Wash contaminated clothing before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : CMIT/MIT mixture