missing translation for 'onlineSavingsMsg'
Learn More

BD FITC BrdU Flow Kit

Product Code. 15819098
Click to view available options
No. of Tests:
50
200
Unit Size:
1 set
Each
This item is not returnable. View return policy

Product Code. 15819098

Brand: BD 557891

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

One 65μL vial fluorochrome-conjugated Anti-BrdU antibody, one 25mL bottle BD Cytofix/Cytoperm™ buffer, two 25mL bottles BD perm/wash™ buffer (10X), one 10mL bottle BD Cytoperm™, DNase; BrdU; 7-AAD; Kit Manual

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
Includes One 65μL vial fluorochrome-conjugated Anti-BrdU antibody, one 25mL bottle BD Cytofix/Cytoperm™ buffer, two 25mL bottles BD perm/wash™ buffer (10X), one 10mL bottle BD Cytoperm™
No. of Tests 200
Regulatory Status RUO
Product Type FITC BrdU Flow Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.