missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD FITC BrdU Flow Kit
Click to view available options
No. of Tests:
50
200
Unit Size:
1 set
Each
Description
Includes
One 65μL vial fluorochrome-conjugated Anti-BrdU antibody, one 25mL bottle BD Cytofix/Cytoperm™ buffer, two 25mL bottles BD perm/wash™ buffer (10X), one 10mL bottle BD Cytoperm™, DNase; BrdU; 7-AAD; Kit Manual
Specifications
Specifications
| Content And Storage | 4°C |
| Includes | One 65μL vial fluorochrome-conjugated Anti-BrdU antibody, one 25mL bottle BD Cytofix/Cytoperm™ buffer, two 25mL bottles BD perm/wash™ buffer (10X), one 10mL bottle BD Cytoperm™ |
| No. of Tests | 200 |
| Regulatory Status | RUO |
| Product Type | FITC BrdU Flow Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction