missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD FoxP3 Staining Kit
High performance reagent kit for detection of human FoxP3 positive regulatory T cells
Brand: BD 560132
Includes: PE/Alexa Fluor™ 647/ Alexa Fluor 488, FoxP3, CD4, CD25, Buffer set
Description
- Complementing antibody conjugates buffer system fixes and permeabilizes cells in easy, simple steps
- Components are sufficient for 100 tests
- Clone: M-A251, RPA-T4, 259D/C7
- Isotype: mouse IgG1, IgG1, κ, IgG1, κ
Flow Cytometry
Specifications
Alexa Fluor 647 | |
1 Kit |
Research use only