Learn More
BD GasPak™ EZ Gas Generating Systems Incubation Containers
Produce atmospheres suitable to support the primary isolation and cultivation of a wide range of bacteria. BD Difco™ GasPak™ EZ Gas Generating Systems Incubation Containers are multi-use systems able to support the primary isolation and cultivation of anaerobic, microaerophilic, or capnophilic bacteria by use of gas generating sachets inside multi-use incubation containers.
Brand:  BD 260671
 View more versions of this product
Product Code. 10118924
  • £864.00 / Each
Estimated Shipment: 03-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Produce atmospheres suitable to support the primary isolation and cultivation of a wide range of bacteria. BD Difco™ GasPak™ EZ Gas Generating Systems Incubation Containers are multi-use systems able to support the primary isolation and cultivation of anaerobic, microaerophilic, or capnophilic bacteria by use of gas generating sachets inside multi-use incubation containers.
  • Non-breakable, chemical resistant containers
  • Removable Petri dish racks are available (sold separately)
  •  
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EZ Incubation Container
2°C to 25°C
Complete Box
Petri Dishes (15 to 18)
Each
Easy close latches
Reusable
Primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets
GasPak
Standard
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
BD GasPak™ EZ Gas Generating Systems Incubation Containers > Standard Incubation Container