missing translation for 'onlineSavingsMsg'
Learn More

BD GasPak™ EZ Gas Generating Systems and Supplies: Container System Sachets

Used in producing anaerobic, microaerophilic, or CO2-enriched environments.   BD Difco™ GasPak™ EZ Gas Generating Systems and Supplies: Container System Sachets provide catalyst-free convenience.

Brand:  BD 260679

Code : 04

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 11757194

  • £108.00 / Pack of 20
Estimated Shipment: 04-09-2024
to see stock.
Add to basket

Description

Description

  • For use in both the GasPak™ EZ Containers and the GasPak™ 100 and GasPak™ 150 jars
  • Sachets require no water or catalyst for activation and are engineered to provide rapid gas generation for optimal bacterial growth

Specifications

Specifications

EZ CO2 Container System Sachet
Sachet
EZ CO2 container system
GasPak
2°C to 25°C
Primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets
20 Sachets/Box
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Catalyst-free convenience for use in producing anaerobic, microaerophilic, or CO2-enriched environments