missing translation for 'onlineSavingsMsg'
Learn More

BD GasPak™ EZ Gas Generating Systems and Supplies: Pouch Systems

Used in producing anaerobic, microaerophilic, or CO2-enriched environments. BD Difco™ GasPak™ EZ Gas Generating Systems and Supplies provide catalyst-free convenience.

£131.00 - £328.00

Specifications

Closure Type Ziplock
Format Resealable Pouches
For Use With (Application) Primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets
For Use With (Equipment) BD GasPak EZ Small Incubation Container
Product Line GasPak
View More Specs

Products 3
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
11777194
View accessory products View Documents
BD
260683
EZ Anaerobe Pouch System
£328.00
Pack of 20
Please to purchase this item. Need a web account? Register with us today!
11787194
View accessory products View Documents
BD
260684
EZ CO2 Pouch System
£132.00
Pack of 20
Please to purchase this item. Need a web account? Register with us today!
11797194
View accessory products View Documents
BD
260685
EZ CampyPouch System
£131.00
Pack of 20
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • One-step resealable pouches
  • The pouches are specially designed to maximize the preservation of the desired environment throughout incubation
  • This heat-free technology minimizes condensate, allowing excellent bacterial colony visualization though the pouch
Specifications

Specifications

Ziplock
Primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets
GasPak
Resealable Pouches
BD GasPak EZ Small Incubation Container
20 Sachets
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BD GasPak™ EZ Gas Generating Systems and Supplies: Pouch Systems

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.