missing translation for 'onlineSavingsMsg'
Learn More

BD General Caspase Inhibitor, Z-VAD-FMK

Cell permeable general caspase inhibitor that binds to the catalytic site of caspase proteases irreversibly and inhibits apoptosis

Brand:  BD 550377

Product Code. 15826258

  • £276.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Members of the caspase family play key roles in inflammation and mammalian apoptosis. The Z-VAD-FMK peptide is O-methylated in the P1 position on aspartic acid providing enhanced stability and increased cell permeability. The inhibitor is designed to be used in both in vivo and in vitro cell based assays to measure the inhibition of apoptosis.

  • Molecular weight: 467 Daltons
  • Form: Lyophilized
  • Storage buffer: Lyophilized in dimethyl sulfoxide (DMSO)

Avoid multiple freeze-thaws of product.

Flow cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Flow cytometry
-20°C
467
Lyophilized
Lyophilized in Dimethyl Sulfoxide
1 mg
Z-VAD-FMK
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For research use only.