missing translation for 'onlineSavingsMsg'
Learn More

BD HeLa + Pervanadate Lysate

Useful control for the western blotting application

Brand:  BD 612015

Additional Details : Weight : 0.01000kg

Product Code. 15886069

  • £775.00 / Each
Estimated Shipment: 22-08-2024
to see stock.

Includes: Human Epitheloid Cervical Carcinoma

Description

Description

  • Rich in endogenously expressed proteins
  • Convenient to use and provided in a denatured format allowing for immediate loading onto an SDS-PAGE gel

Specifications

Specifications

Western Blot
HeLa (Human Epitheloid Carcinoma) cells, were grown to 80 to 90% confluency, then treated with 1mM pervanadate, a general inhibitor of Protein Tyrosine Phosphatases, for 15 min. at 37°C
1mg/mL in SDS-PAGE Buffer: 62mM Tris pH 6.8, 2% SDS, 0.9% beta-mercaptoethanol, 0.003% Bromophenol Blue, 5% Glycerol
1 mg/mL
Epitheloid Cervical Carcinoma
500 μg
Human
-20°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only