missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Hoechst 33342 Solution
Click to view available options
Quantity:
1 mg
Unit Size:
Each
Description
Hoechst 33342 is a bisbenzimidazole dye with high specificity for binding to double-stranded DNA (preferentially binds to A-T base pairs). This dye is useful to label double-stranded DNA and thus to visualize nuclei.
- Can be excited at ∽355nm by UV light source (UV laser beam or mercury arc-lamp)
- Emits blue fluorescence light around emission maximum at 461 nm when bound to DNA
- Ribonuclease treatment is not needed to avoid nonspecific RNA staining
- Concentration: 1mg/mL
- Size: 1mg
Bioimaging/Immunofluorescence
Specifications
Specifications
| Product Type | Hoechst 33342 solution |
| Quantity | 1 mg |
| For Use With (Application) | Fluorescent Staining, Bioimaging, Immunofluorescence (IF) |
| Content And Storage | 4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction