Learn More
BD CFSE
Used for flow cytometric monitoring of cell divisions. BD Horizon™ CFSE is non-fluorescent dye passively diffuses across cell membranes and is cleaved by intracellular esterases within viable cells.
Brand:  BD 565082
Product Code. 15864069
  • £146.00 / Each
Estimated Shipment: 02-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Blue laser excitable dye
  • Used for flow cytometric monitoring of cell divisions
  • The non-fluorescent dye passively diffuses across cell membranes and is cleaved by intracellular esterases within viable cells
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1mg
Blue
Laser excitable dye
Intracellular staining (flow cytometry)
≤ -20°C