missing translation for 'onlineSavingsMsg'
Learn More

BD BV480 Mouse Anti-Human CD138

CD138 is strongly expressed on multiple-myeloma-derived cell lines and malignant plasma cell populations. BD Horizon™ BV480 Mouse Anti-Human CD138 binds to many extracellular matrix proteins through its heparan sulfate side-chains, like fibronectin, collagen types I, III, and V, tenascin, thrombospondin, and antithrombin III.

Brand:  BD 566140

Product Code. 15854779

  • £635.00 / Each
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
  • BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BV480 Mouse Anti-Human CD138
Flow Cytometry
BV480
Store undiluted at 4°C
100 Tests
QC Testing: Human
5 μL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers