missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BV480 Mouse Anti-Human IgD
mIgD is expressed on mature naive B cells (along with membrane IgM) and serves as a B-cell receptor for antigen (BCR). BD Horizon™ BV480 Mouse Anti-Human IgD BCR, in association with other signaling molecules including CD79a and CD79b, can transduce activating or tolerizing signals intracellularly into B lymphocytes.
£299.00 - £565.00
Specifications
Product Type | BV480 Mouse Anti-Human IgD |
---|---|
For Use With (Application) | Flow Cytometry |
Cross Reactivity | QC Testing: Human |
Form | BV480 |
Sample Volume | 5 μL |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15814829
|
BD
566187 |
25 Tests |
£299.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
15834779
|
BD
566138 |
100 Tests |
£565.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
Description
- With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
- BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications
BV480 Mouse Anti-Human IgD | |
QC Testing: Human | |
5 μL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide |
Flow Cytometry | |
BV480 | |
Store undiluted at 4°C |