missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BV480 Mouse Anti-Human TCR γδ
The γδ TCR is associated with the signal-transducing CD3 complex. BD Horizon™ BV480 Mouse Anti-Human TCR γδ is expressed by thymocytes and by peripheral T cell subsets (γδ T cells) that are located in the blood, liver, skin and various lymphoid and mucosal tissues. γδ T cells contribute to both innate and adaptive immune responses to infections and tumors.
£301.00 - £620.00
Specifications
Product Type | BV480 Mouse Anti-Human TCR γδ |
---|---|
For Use With (Application) | Flow Cytometry |
Concentration | 0.2 mg/mL |
Cross Reactivity | QC Testing: Human |
Form | BV480 |
Description
- With an Ex Max of 436nm and Em Max at 478nm, BD Horizon BV480 can be excited by the violet laser and detected in the BD Horizon BV510 (525/40nm) filter set
- BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications
BV480 Mouse Anti-Human TCR γδ | |
0.2 mg/mL | |
BV480 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide |
Flow Cytometry | |
QC Testing: Human | |
Store undiluted at 4°C |