Learn More
BD BV480 Rat Anti-Mouse CD19
Expressed throughout B-lymphocyte development from the pro-B cell through the mature B-cell stages. BD Horizon™ BV480 Rat Anti-Mouse CD19 participates in B-lymphocyte development, B-cell activation, maturation of memory B cells and regulation of tolerance. CD19 has also been detected on peritoneal mast cells, co-localized with CD21/CD35, and it is proposed to play a role in complement-mediated mast-cell activation.
Brand: BD 566107
Additional Details : Weight : 0.01000kg
Description
- With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
- BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications
BV480 Rat Anti-Mouse CD19 | |
Flow Cytometry | |
QC Testing: Mouse | |
Store undiluted at 4°C |
0.1 mg | |
0.2 mg/mL | |
BV480 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide |