missing translation for 'onlineSavingsMsg'
Learn More

BD Human Endothelial Cell Lysate

Cell or tissue lysates are a useful control for the western blotting application

Brand:  BD 611450

Additional Details : Weight : 0.01000kg

Product Code. 15805939

  • £1345.00 / Each
Estimated Shipment: 22-08-2024
to see stock.

Includes: Human Endothelium

Description

Description

A source that can be rich in endogenously expressed proteins, this lysate is convenient to use and provided in a denatured format allowing for immediate loading onto an SDS-PAGE gel.

  • Storage buffer: SDS-PAGE buffer (62mM Tris pH 6.8, 2% SDS, 0.9% b-mercaptoethanol, 0.003% bromophenol blue, 5% glycerol)

Western blot

Specifications

Specifications

Western Blot
500 μg
Human
-20°C
Endothelial Cells
1mg/mL in SDS-PAGE Buffer: 62mM Tris pH 6.8, 2% SDS, 0.9% beta-mercaptoethanol, 0.003% Bromophenol Blue, 5% Glycerol
1 mg/mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only