missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Human MCP-1 ELISA Kit
Quantitative determination of human MCP-1 in serum, plasma and cell culture supernatant
Brand: BD 559017
Additional Details : Weight : 0.25000kg
Includes: 2 plates of 96 breakable wells coated with anti-human MCP-1 antibody; Detection antibody: Biotinylated anti-human MCP-1; Enzyme Concentrate (250X): Horseradish peroxidase conjugate; Recombinant human MCP-1, lyophilized, 4 vials; Standard Diluent; Wash Concentrate (20X); Substrate Reagent A; Substrate Reagent B; Stop Solution; Adhesive Plate Sealers
Description
- Solid phase sandwich ELISA
- Utilizes monoclonal antibody specific for MCP-1 coated on a 96-well plate
Applications:
ELISA
Specifications
MCP-1 | |
2 Plates of 96 breakable well coated with Anti-Human MCP-1 Antibody, Biotinylated Anti-Human MCP-1, HRP Conjugate (250X), 4 vials recombinant Human MCP-1 (lyophilized), standard Diluent, wash concentrate (20X), Substrate reagent A, Substrate reagent B, stop solution, adhesive plate sealers | |
Serum, Plasma, Cell Culture Supernatant | |
Quantitative determination of Human MCP-1, ELISA |
Human | |
2 Plates | |
2°C to 8°C | |
Human MCP-1 ELISA kit |
Research use only