missing translation for 'onlineSavingsMsg'
Learn More

BD Biosciences™ BD IntraSure™ Kit (CE/IVD)

Intended to fix and permeabilize cells to enable the staining of intracellular targets with minimal effect on the prior staining of cell surface antigens.

Brand:  BD Biosciences™ 641778

Additional Details : Weight : 0.07800kg


Product Code. 15846299

  • £195.00 / Each
Estimated Shipment: 29-07-2024
to see stock.

Description

Description

  • The BD IntraSure™ kit is intended to fix and permeabilize cells to enable the staining of intracellular targets with minimal effect on the prior staining of cell surface antigens.
  • Samples are acquired using a BD FACS™ flow cytometer equipped with the appropriate analysis software for data acquisition and analysis.
Specifications

Specifications

5 mL of Reagent A, a proprietary buffered fixative containing 1% formaldehyde, and 2.5 mL of Reagent B, a proprietary buffered permeabilization solution containing a detergent
Store bottles at room temperature (20°C to 25°C) and protect from light. Do not freeze.
Each reagent is stable until the expiration date shown on the bottle label when stored as directed.
50 Tests
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • BD IntraSure Kit CE-IVD

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : formaldehyde, methanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers