Learn More
BD Macro-Vue™ RPR Cards and Components
For large-scale testing in public health and clinical laboratories
£197.00
Specifications
For Use With (Application) For large-scale testing in public health and clinical laboratories
View More Specs
Products 1
Product Code Brand Quantity Type Price Quantity & Availability  
Product Code Brand Quantity Type Price Quantity & Availability  
17352421
View Documents
BD
276709
10 Pack RPR Control Cards
£197.00
Pack of 10
Description
Description
RPR Card antigen suspension is a carbon particle cardiolipin antigen1 which detects reagin, an antibody-like substance present in serum or plasma from syphilitic persons, and occasionally in serum or plasma of persons with other acute or chronic conditions. The reagin binds to the test antigen, which consists of cardiolipin-lecithin-coated cholesterol particles, causing macroscopic flocculation.
  • CLIA Complexity: Moderate
  • Nontreponemal test
  • For the serological detection of syphilis as described in A Manual of Tests for Syphilis
Specifications
Specifications
For large-scale testing in public health and clinical laboratories
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
BD Macro-Vue™ RPR Cards and Components